MilliporeSigma
Immobilization of proteins on agarose beads, monitored in real time by bead injection spectroscopy.
The Analyst (2006-06-28)
Jaromir Ruzicka, Andrea D Carroll, Ilkka Lähdesmäki
RÉSUMÉ
A novel approach to real-time monitoring of protein immobilization resulted in the surprising finding that current immobilization protocols are far from optimized.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
IgG from human serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution