MilliporeSigma
The upregulation of annexin A2 after spinal cord injury in rats may have implication for astrocyte proliferation.
Neuropeptides (2016-11-12)
Jiajia Chen, Zhiming Cui, Saishuai Yang, Chunshuai Wu, Weidong Li, Guofeng Bao, Guanhua Xu, Yuyu Sun, Lingling Wang, Jinlong Zhang
RÉSUMÉ
Annexin A2 (ANXA2), is a member of the annexin family of proteins that exhibit Ca
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human ANXA2