MilliporeSigma
  • Selective κ receptor partial agonist HS666 produces potent antinociception without inducing aversion after i.c.v. administration in mice.
Selective κ receptor partial agonist HS666 produces potent antinociception without inducing aversion after i.c.v. administration in mice.
British journal of pharmacology (2017-05-12)
Mariana Spetea, Shainnel O Eans, Michelle L Ganno, Aquilino Lantero, Michael Mairegger, Lawrence Toll, Helmut Schmidhammer, Jay P McLaughlin
RÉSUMÉ
The κ receptor has a central role in modulating neurotransmission in central and peripheral neuronal circuits that subserve pain and other behavioural responses. Although κ receptor agonists do not produce euphoria or lead to respiratory suppression, they induce dysphoria and sedation. We hypothesized that brain-penetrant κ receptor ligands possessing biased agonism towards G protein signalling over β-arrestin2 recruitment would produce robust antinociception with fewer associated liabilities. Two new diphenethylamines with high κ receptor selectivity, HS665 and HS666, were assessed following i.c.v. administration in mouse assays of antinociception with the 55°C warm-water tail withdrawal test, locomotor activity in the rotorod and conditioned place preference. The [ HS665 (κ receptor agonist) and HS666 (κ receptor partial agonist) demonstrated dose-dependent antinociception after i.c.v. administration mediated by the κ receptor. These highly selective κ receptor ligands displayed varying biased signalling towards G protein coupling in vitro, consistent with a reduced liability profile, reflected by reduced sedation and absence of conditioned place aversion for HS666. HS665 and HS666 activate central κ receptors to produce potent antinociception, with HS666 displaying pharmacological characteristics of a κ receptor analgesic with reduced liability for aversive effects correlating with its low efficacy in the β-arrestin2 signalling pathway. Our data provide further understanding of the contribution of central κ receptors in pain suppression, and the prospect of dissociating the antinociceptive effects of HS665 and HS666 from κ receptor-mediated adverse effects.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)