MilliporeSigma
Metadherin promotes metastasis by supporting putative cancer stem cell properties and epithelial plasticity in pancreatic cancer.
Oncotarget (2017-10-17)
Kensuke Suzuki, Shigetsugu Takano, Hideyuki Yoshitomi, Hitoe Nishino, Shingo Kagawa, Hiroaki Shimizu, Katsunori Furukawa, Masaru Miyazaki, Masayuki Ohtsuka
RÉSUMÉ
Pancreatic ductal adenocarcinoma (PDAC) has a high metastatic potential. However, the mechanism of metastatic colonization in PDAC remains poorly understood. Metadherin (MTDH) has emerged in recent years as a crucial mediator of metastasis in several cancer types, although the biological role of MTDH in PDAC has not been investigated. Here, we demonstrated the functional roles of MTDH in PDAC progression, especially focusing on the metastatic cascade.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human MTDH