MilliporeSigma
CSF-1R regulates non-small cell lung cancer cells dissemination through Wnt3a signaling.
American journal of cancer research (2017-12-09)
Yan Xia Yu, Hai Jian Wu, Bing Xu Tan, Chen Qiu, Hui Zhong Liu
RÉSUMÉ
Therapeutic antibodies targeting colony stimulating factor 1 receptor (CSF-1R) to block colony stimulating factor-1/colony stimulating factor 1 receptor (CSF-1/CSF-R) signaling axis have exhibit remarkable efficacy in the treatment of malignant tumor. Yet, little is known about the effects of intrinsic CSF-1R in human non-small-cell carcinoma (NSCLC). Here we demonstrated that NSCLC cell-intrinsic CSF-1R promoted cells growth and metastasis both in vitro and in vivo. CSF-1R knocked-down by transfecting with shRNA target CSF-1R suppressed NSCLC cells proliferation and tumor growth in nude mice. Conversely, ectopic expression of CSF-1R promoted cells proliferation and accelerated tumor growth. Mechanistically, the NSCLC CSF-1R modulated downstream effectors of phosphatidylinositol 3-kinase (PI3K) signaling. In addition, CSF-1R overexpression significantly enhanced NSCLC cells mobility, invasion and epithelial-mesenchymal transition (EMT) process, whereas silencing CSF-1R inhibits these phenotypes. Microarray analysis suggested that Wnt family member 3a (Wnt3a) function as a downstream factor of CSF-1R. On account of this, we future identified CSF-1R/Wnt3a a signaling pathway sustained NSCLC cells metastasis. Finally, in patients, CSF-1R and Wnt3a expression positively correlated with the of NSCLC patients. Our results identify NSCLC cell intrinsic functions of CSF-1R/Wnt3a axis in dissemination of NSCLC.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Wnt3a