MilliporeSigma
A new one-Pot synthesis of alkynylphosphonates
Organic letters (2000-12-02)
Lera, Hayes
RÉSUMÉ
A method for the palladium-catalyzed synthesis of alkynylphosphonates from 1,1-dibromo-1-alkenes has been developed. In general, the best catalyst system for this transformation was found to be Pd(OAc)(2), dppf, H-phosphonate, propylene oxide, DMF, 80 degrees C. The reaction appears tolerant of a range of functional groups in both the 1,1-dibromo-1-alkene and H-phosphonate coupling partners. The synthesis of a backbone-modified thymidine dimer is used to illustrate the application of this methodology in the synthesis of complex target molecules.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
DPPF, 97%