MilliporeSigma
Oncolytic Virotherapy Blockade by Microglia and Macrophages Requires STAT1/3.
Cancer research (2017-11-10)
Zahid M Delwar, Yvonne Kuo, Yan H Wen, Paul S Rennie, William Jia
RÉSUMÉ
The first oncolytic virotherapy employing HSV-1 (oHSV-1) was approved recently by the FDA to treat cancer, but further improvements in efficacy are needed to eradicate challenging refractory tumors, such as glioblastomas (GBM). Microglia/macrophages comprising approximately 40% of a GBM tumor may limit virotherapeutic efficacy. Here, we show these cells suppress oHSV-1 growth in gliomas by internalizing the virus through phagocytosis. Internalized virus remained capable of expressing reporter genes while viral replication was blocked. Macrophage/microglia formed a nonpermissive OV barrier, preventing dissemination of oHSV-1 in the glioma mass. The deficiency in viral replication in microglial cells was associated with silencing of particular viral genes. Phosphorylation of STAT1/3 was determined to be responsible for suppressing oHSV-1 replication in macrophages/microglia. Treatment with the oxindole/imidazole derivative C16 rescued oHSV-1 replication in microglia/macrophages by inhibiting STAT1/3 activity. In the U87 xenograft model of GBM, C16 treatment overcame the microglia/macrophage barrier, thereby facilitating tumor regression without causing a spread of the virus to normal organs. Collectively, our results suggest a strategy to relieve a STAT1/3-dependent therapeutic barrier and enhance oHSV-1 oncolytic activity in GBM.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Aminoguanidine hydrochloride, ≥98%