Größe auswählen
CHF 469.00
CHF 469.00
Über diesen Artikel
DE-M marking
with recirculation valve
GL 32
GL 38
S 40
Fortfahren mit
Produktname
BRAND® Dispensette® S Digital bottle-top dispenser, volume range 0.2-2 mL, with recirculation valve
feature
incremental 0.01 mL
DE-M marking
with recirculation valve
manufacturer/tradename
BRAND 4600321
volume range
0.2-2 mL
fitting
(GL 28/S28)
GL 32
GL 38
S 40
Verwandte Kategorien
1 of 4
Dieser Artikel | BR4600331 | BR4600351 | BR4600341 |
|---|---|---|---|
| manufacturer/tradename BRAND 4600321 | manufacturer/tradename BRAND 4600331 | manufacturer/tradename BRAND 4600351 | manufacturer/tradename BRAND 4600341 |
| feature incremental 0.01 mL, with recirculation valve, DE-M marking | feature incremental 0.02 mL, DE-M marking, with recirculation valve | feature incremental 0.1 mL, DE-M marking, with recirculation valve | feature incremental 0.05 mL, DE-M marking, with recirculation valve |
| fitting (GL 28/S28), GL 38, GL 32, S 40 | fitting (GL 28/S28), GL 32, GL 38, S 40 | fitting (GL 32-33), GL 38, S 40 | fitting (GL 28/S28), GL 32, GL 38, S 40 |
| volume range 0.2-2 mL | volume range 0.5-5 mL | volume range 2.5-25 mL | volume range 1-10 mL |
Features and Benefits
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
General description
Other Notes
Legal Information
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
Protokolle
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Verwandter Inhalt
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.