Merck
BR4630351
BRAND® Dispensette® S Organic, Digital bottle-top dispenser
volume range 2.5-25 mL, with recirculation valve
Synonym(e):
bottle top dispensers
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
1 EA
CHF 582.00
CHF 582.00
Warenkorb auf Verfügbarkeit prüfen
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
BRAND 4630351
Feature:
incremental 0.1 mL, DE-M marking, with recirculation valve
Fitting:
(GL 32-33), GL 38, S 40
Fortfahren mit
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
feature
incremental 0.1 mL, DE-M marking, with recirculation valve
manufacturer/tradename
BRAND 4630351
volume range
2.5-25 mL
fitting
(GL 32-33), GL 38, S 40
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 4
Dieser Artikel
BR4630361BR4630341BR4630331
manufacturer/tradename
BRAND 4630351
manufacturer/tradename
BRAND 4630361
manufacturer/tradename
BRAND 4630341
manufacturer/tradename
BRAND 4630331
feature
incremental 0.1 mL, with recirculation valve, DE-M marking
feature
incremental 0.2 mL, DE-M marking, with recirculation valve
feature
incremental 0.05 mL, DE-M marking, with recirculation valve
feature
incremental 0.02 mL, DE-M marking, with recirculation valve
fitting
(GL 32-33), S 40, GL 38
fitting
(GL 32-33), GL 38, S 40
fitting
(GL 28/S 28), GL 32, GL 38, S 40
fitting
(GL 28/S 28), GL 32, GL 38, S 40
volume range
2.5-25 mL
volume range
5-50 mL
volume range
1-10 mL
volume range
0.5-5 mL
General description
Dispensette S Organic is ideal for dispensing of organic solvents and acids
Features and Benefits
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Other Notes
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Protokolle
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Verwandter Inhalt
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.