Merck
All Photos(1)

1.18343

Supelco

Calibration standard 3000 T

turbidity 10000 NTU±200

Synonym(s):
Kalibrationsstandard 3000 T

technique(s)

photometry: suitable

Trübung

10000 NTU±200

Kompatibilität

for use with Turbiquant

Lagertemp.

15-25°C

Allgemeine Beschreibung

10 000 NTU Turbiquant
The Turbiquant Calibration Standard is suitable to calibrate the Turbiquant 3000 T. The standard offers the following advantages:
1. The new primary standards have approvals which secondary standards do not have (rated to be a primary standard according to US EPA).
2. Instruments used in drinking water analysis no longer need to be certified with separate primary standards.
3. Clear records of final inspection are included in each set.
4. The new standards have an unrivalled precision due to production accuracy, and stability in solution.
5. The standards are ready-to-use.
6. They are non-toxic.
7. They are unproblematic to store and transport.

Verpackung

1, 1 unit

Hinweis zur Analyse

Messwert (Standard): 10000 FNU ±2 %

Rechtliche Hinweise

TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany

Lagerklassenschlüssel

10-13 - German Storage Class 10 to 13

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service