Direkt zum Inhalt
Merck

CRM-ASP-MUS-D

Domoic Acid Mussel Tissue CRM

certified reference material ()

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern

Über diesen Artikel

UNSPSC Code:
41116107
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


grade

certified reference material ()

agency

National Research Council of Canada (NRC)

description

Analyte group-Algal toxins

manufacturer/tradename

National Research Council of Canada (NRC)

application(s)

food and beverages

storage temp.

−70°C

General description

CRM-ASP-Mus-d is a mussel tissue matrix CRM containing DA and some of its isomers. It is intended to test the accuracy of entire analytical methods or to assist in the development of new analytical methods. For more information, please refer to the Certificate of Analysis or Safety Data Sheet. https://doi.org/10.4224/crm.2011.atx.20100721

Features and Benefits

NRC Canada’s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034. For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis. With NRC Canada’s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory’s accuracy and credibility with NRC Canada’s certified reference materials.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.




Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen



Global Trade Item Number

SKUGTIN
CRM-ASP-MUS-D04065270773036