Merck
All Photos(1)

PE1194

Supelco

Absetzbare Stoffe - WP

Proficiency Testing Material, Concentrate for dilution to 1L

EC-Nummer:
NACRES:
NA.24

Qualitätsniveau

Verpackung

pkg of of × 1EA

Matrix

silicon dioxide

application(s)

environmental

Format

matrix material

Compare Similar Items

View Full Comparison

1 of 4

This Item
QC1194SQC082ERMFD100
Supelco

Supelco

PE1194

Absetzbare Stoffe - WP

Supelco

Supelco

QC1194

Absetzbare Stoffe - WP

Supelco

Supelco

SQC082

Feststoffe in Erde

vibrant-m

ERMFD100

Kieselsol

matrix

silicon dioxide

matrix

NaCl

matrix

soil

matrix

-

application(s)

environmental

application(s)

environmental

application(s)

environmental

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

format

matrix material

format

matrix material

format

matrix material

packaging

pkg of of × 1EA

packaging

jar of 30 mL

packaging

-

packaging

-

Quality Level

100

Quality Level

300

Quality Level

300

Quality Level

-

Allgemeine Beschreibung

Settleable Solids - WP proficiency testing material is produced in accordance with ISO/IEC 17043:2010. Also referred to as non-Potable water or Water Pollution sample, this sample is produced according to TNI requirements. However, please note, this sample may include non-TNI analytes as well. The higher criteria from the TNI tables mimic the levels common to wastewater samples. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Most water PT samples are available as concentrates or ready-to-use volumes giving the end-user flexibility in selecting samples that will fit their exact needs. Our WP program is designed to meet the quality assurance requirements in compliance with Discharge Monitoring Report - Quality Assurance Program (DMR-QA), thereby ensuring the analytical competence of your laboratory.

Type of PT Scheme: Water analysis - Water Pollution (WP)

Matrix: Water Pollution/Wastewater

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Settleable Solids - WP can be found in the reporting packet which will be available to download at PT Portal.

Anwendung

Settleable Solids - WP is a non-Potable water (NPW) PT standard, suitable for testing settleable solids in water pollution/wastewater.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Leistungsmerkmale und Vorteile

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Angaben zur Herstellung

Die Werte der Analyten sind Lot-spezifisch.

Lagerung und Haltbarkeit

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Sonstige Hinweise

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2022-2023)

Wastewater
WP22-1 January 12 – February 25, 2022
WP22-2 March 23 - May 6, 2022
WP22-3 July 6 - August 19, 2022
WP22-4 September21 - November 4, 2022

WP23-1 January 11 – February 24, 2023
WP23-2 March 22 – May 5, 2023
WP23-3 July 5 – August 18, 2023
WP23-4 September 20 – November 3, 2023

DMR-QA (US customers only)
DMR-QA 43 2023 Dates TBD

Quick-Turn Study (on demand proficiency test that is offereddepending on lot availability)

Rechtliche Hinweise

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Settleable solids 5-50 mL/L

Piktogramme

Health hazard

Signalwort

Danger

H-Sätze

Gefahreneinstufungen

STOT RE 1 Inhalation

Zielorgane

Lungs

Lagerklassenschlüssel

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flammpunkt F

Not applicable

Flammpunkt C

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service