CLS430588
Corning® non-treated culture dishes
D × H 35 mm × 10 mm
Synonym(e):
cell culture dishes, cell culture plates, culture dish
Größe auswählen
CHF 270.00
Größe auswählen
About This Item
Fortfahren mit
CHF 270.00
Materialien
polystyrene
round
Sterilität
sterile
Verwendung
1.6-2.4 mL (Optimal culture medium volume)
Verpackung
sleeve of 20
Hersteller/Markenname
Corning 430588
T × H
35 mm × 10 mm
Oberflächenbereich
8 cm2
1 of 4
Dieser Artikel | CLS430589 | CLS430591 | CLS430167 |
---|---|---|---|
material polystyrene , round | material polystyrene , round | material polystyrene , round | material polystyrene , round |
sterility sterile | sterility sterile | sterility sterile | sterility sterile |
manufacturer/tradename Corning 430588 | manufacturer/tradename Corning 430589 | manufacturer/tradename Corning 430591 | manufacturer/tradename Corning 430167 |
packaging sleeve of 20 | packaging sleeve of 20 | packaging sleeve of 20 | packaging sleeve of 20 |
usage 1.6-2.4 mL (Optimal culture medium volume) | usage - | usage - | usage 11.0-16.5 mL (Optimal culture medium volume) |
D × H 35 mm × 10 mm | D × H 60 mm × 15 mm | D × H 100 mm × 20 mm | D × H 100 mm × 20 mm |
Allgemeine Beschreibung
- Manufactured from optically-clear virgin polystyrene
- Not treated for cell attachment, for applications where cell attachment is not desired
- Sterilized by gamma radiation and certified nonpyrogenic
- Have stacking beads to aid in handling
- Supplied with vents to provide consistent gas exchange
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)