Merck
TMO140156
Nunc® OmniTray Single-Well Plate With Lid
Nunclon Delta surface, without notched corners, polystyrene, clear, sterile, case of 90 ea
Synonym(e):
microplate, treated microplate, Nunc 140156, cultivation plate, culture dish, Nunc® OmniTray(TM) Single-Well Plate un-notched
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
90 EA
CHF 673.00
CHF 673.00
Warenkorb auf Verfügbarkeit prüfen
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41122102
Material:
clear , polystyrene , rectangular wells
Sterility:
sterile
Binding type:
Nunclon Delta surface
Manufacturer/tradename:
Nunc 140156
Packaging:
case of 90 ea
Fortfahren mit
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
clear , polystyrene , rectangular wells
sterility
sterile
feature
lid, skirt: no
packaging
case of 90 ea
manufacturer/tradename
Nunc 140156
L × W
128 mm × 86 mm
capacity
90 mL
well volume
90 mL
well working volume
35 mL
working volume
35 mL
suitability
suitable for (standard petri dish techniques and bacterial growth, screening libraries and DNA hybridization methods)
binding type
Nunclon Delta surface
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 4
Dieser Artikel
TMO165218O0764TMO150787
manufacturer/tradename
Nunc 140156
manufacturer/tradename
Nunc 165218
manufacturer/tradename
Nunc 242811
manufacturer/tradename
Nunc 150787
sterility
sterile
sterility
sterile
sterility
sterile
sterility
-
material
clear , polystyrene
material
clear , polystyrene
material
polystyrene
material
polystyrene
packaging
case of 90 ea
packaging
case of 60 ea
packaging
pack of 10, case of 60
packaging
case of 75 ea
General description
Made from clear polystyrene with standard microplate footprint, the Thermo Scientific Nunc OmniTrays are ideal for screen libraries and DNA hybridization. Available both as a standard version with 2 notched corners and as a rectangular version optimized for automation. Surface treated for cell culture or non-treated for bacterial growth and colony isolation.
Made from clear polystyrene with standard microplate footprint, the Thermo Scientific™ Nunc™ OmniTrays™ are ideal for screen libraries and DNA hybridization. Available both as a standard version with 2 notched corners and as a rectangular version optimized for automation. Surface treated for cell culture or non-treated for bacterial growth and colony isolation.
Features and Benefits
  • Clear polystyrene with excellent optical properties.
  • External dimensions: 128 x 86mm.
  • Tray can be frozen at -20°C.
  • Cat. No. 140156 (TMO140156) without the notched corners is ideal for automation picking and viewing systems.
  • Nunclon® Delta cell culture-treated surface promotes cell attachment and growth.
  • The Non-treated surface is suitable for bacterial culture.
  • Clear polystyrene with excellent optical properties.
  • External dimensions: 128 x 86mm.
  • Tray can be frozen at -20°C.
  • Cat. No. 140156 without the notched corners is ideal for automation picking and viewing systems.
  • Nunclon™ Delta cell culture-treated surface promotes cell attachment and growth.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)