HomeWebinarsDuolink® PLA Technology: How to detect and quantify protein
Duolink® PLA Technology: How to detect and quantify protein
Webinar
What Does it Cover?
Duolink® proximity ligation assay (PLA) technology allows you to visualize protein interactions with cellular localization and quantities by amplifying signals corresponding to single and post-translational protein events. With 1000x sensitivity and high specificity, this protein detection technology allows to visualize protein functions, all within a native cell. The PLA method provides:
  • Visual protein interactions
    − Both stable and transient
  • Endogenous protein detection
    − No overexpression or genetic manipulation
  • High specificity
    − Use of two antibodies/probes eliminates false positives
  • Single molecule sensitivity
    − Rolling circle amplification makes proteins visible
  • No special equipment needed
    − Standard immunofluorescence methods
This webinar will review how to work with PLA technology and provide an overview of their potential along with example applications.
What Will You Learn?
  • How to work with Duolink® PLA technology
  • Overview of most relevant applications using Duolink®
  • Product offering to run an assay with PLA using Immunofluorescence or Flow Cytometry
  • Benefits of working with highly specific and powerful technology to study protein biology
Who Should Attend?
Researchers in the T-Cell, adaptive immunotherapy, and immuno-oncology fields
Speakers
Cláudia Emanuele, Ph.D.
Merck
Field Application Scientist
Claudia earned her BS in Biology from the University of Juiz de Fora, Brazil, and her MS in Physiology from the University of Sao Paulo. She received her Ph.D. in Physiology from the University of São Paulo, with a scholarship for 6 month at the Weizmann Institute, Israel. Following completion of her Ph.D., she joined Merck as a Field Application Scientist for Biology Portfolio focusing on the Latin America Region.
Paola Braga
Merck
Sales Application Specialist
Paola earned her Bachelor degree in Biologic Science with Medical modality from Metodista University and her Master in Administration and Maketing from FAAP. She joined Merck in 2006 as Technical Advisor for Bioscience portfolio and had transitioned to different roles, always focused on Life Science business. In her present position she works as Application Specialist of Biology portfolio.