Merck
All Photos(4)

151890

Sigma-Aldrich

Deuterium oxide

"100%", ≥99.96 atom % D

Synonym(s):
Heavy water, Water-d2
Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.21

Quality Level

isotopic purity

≥99.96 atom % D

form

liquid

technique(s)

NMR: suitable

bp

101.4 °C (lit.)

mp

3.8 °C (lit.)

SMILES string

[2H]O[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Deuterium oxide (D2O) is a deuterated solvent. It is 100% isotopically enriched solvent for NMR (Nuclear Magnetic Resonance). It is widely employed in high resolution NMR studies. Various thermodynamic properties (such as intermolecular vibrational frequencies, energy of the hydrogen bond, free energy, enthalpy and entropy) of liquid deuterium oxide have been evaluated. Ionization constant for D2O (in the range of 5-50°C), pK values (at 25°C) and enthalpy, entropy, heat capacity change (for the dissociation of D2O) have been reported.

Packaging

10×0.75, 10×1 mL in ampule
10, 50 g in glass bottle
125 g in Sure/Seal™
250 g in glass bottle
1 kg in glass bottle

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service