Skip to Content
Merck

CH6H9A56B928

Chem Impex International, Inc.

Fmoc-2-chloro-L-phenylalanine

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
421.88
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

130 - 136 C

Iupac Name

2-chloro-N-[(9H-fluoren-9-ylmethoxy)carbonyl]phenylalanine

Inchi Code

1S/C24H20ClNO4/c25-21-12-6-1-7-15(21)13-22(23(27)28)26-24(29)30-14-20-18-10-4-2-8-16(18)17-9-3-5-11-19(17)20/h1-12,20,22H,13-14H2,(H,26,29)(H,27,28)

InChI key

RNNKPNPLIMFSDY-UHFFFAOYSA-N

storage temp.

Store at 0 - 8 C

Purity

98%

Country of Origin

US

Shipping Temperature

RT

Product Link

https://www.chemimpex.com/products/04096

Melting Point

130 - 136 C

MSDS

https://www.chemimpex.com/products/04096


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service