Skip to Content
Merck

ENAH30469962

Enamine

3-tert-butyl-3-azabicyclo[3.3.1]nonan-9-ol

Synonym(s):

3-tert-butyl-3-azabicyclo[3.3.1]nonan-9-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
197.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H23NO/c1-12(2,3)13-7-9-5-4-6-10(8-13)11(9)14/h9-11,14H,4-8H2,1-3H3

Inchi Key

COOVPSNCROLCRH-UHFFFAOYSA-N

Country of Origin

UA





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library