Skip to Content
Merck

Skip To

1.37166

Non Animal Origin Squalene

greener alternative

EMPROVE® EXPERT

Synonym(s):

Squalene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C30H50
CAS Number:
Molecular Weight:
410.72
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

EMPROVE® EXPERT

Quality Segment

form

liquid

sustainability

Greener Alternative Product

application(s)

liquid formulation
parenterals
pharmaceutical
vaccine development

greener alternative category

storage temp.

2-8°C

InChI

1S/C30H50/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h15-18,23-24H,9-14,19-22H2,1-8H3/b27-17+,28-18+,29-23+,30-24+

InChI key

YYGNTYWPHWGJRM-AAJYLUCBSA-N

General description

Sterile liquid applications demand excipients with the highest standards of quality, purity and reliability. As part of our Emprove Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Application

Squalene is a lipid widely used in vaccine formulations as an adjuvant, where it enhances immune responses and ensures vaccine efficacy. Non Animal Origin Squalene is a suitable alternative to squalene traditionally sourced from shark liver oil. Due to its low microbial and endotoxin limits, Non Animal Origin Squalene Emprove® Expert is especially suitable for high-risk parenteral applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0002131PHR3519867200O
storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

form

liquid

form

-

form

-

form

(neat oil)

Quality Level

500

Quality Level

100

Quality Level

300

Quality Level

-

application(s)

liquid formulation
parenterals
pharmaceutical
vaccine development

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

vaccine development

greener alternative category

DfS-Developed

greener alternative category

-

greener alternative category

-

greener alternative category

Aligned

sustainability

Greener Alternative Product

sustainability

-

sustainability

-

sustainability

Greener Alternative Product


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library