Merck
All Photos(1)

611786

Supelco

Chloroform solution

NMR reference standard, 5% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.

MDL number:
PubChem Substance ID:
NACRES:
NA.24

grade

NMR reference standard
analytical standard

Quality Level

concentration

5% in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

application(s)

environmental

format

single component solution

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

1H line shape

Quantity

3 mm O.D. tube contains 0.235 mL.

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service