All Photos(1)

639141

Sigma-Aldrich

Water solution

contains 0.1 % (v/v) ammonium hydroxide

Linear Formula:
H2O
MDL number:
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

contains

0.1 % (v/v) ammonium hydroxide

technique(s)

HPLC: suitable

application(s)

food and beverages

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The product is water containing 0.1%v/v ammonium hydroxide that can be employed for HPLC and UHPLC (Ultra High performance Liquid Chromatography) techniques. This pre-blended mobile phase solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases.

Packaging

20 L in Nowpak™

Preparation Note

Product filtered through a 0.2 μm filter

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service