Merck
All Photos(1)

59418C

SAFC

Trypsin-EDTA Solution 10X

Synonym(s):
Trypsin solution from porcine pancreas
CAS Number:
MDL number:
NACRES:
NA.75

description

for research or for further manufacturing use

Quality Level

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Trypsin-EDTA Solution 10X has been used to release adherent cells from tissue culture plates for passaging.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Quotes and Ordering

Molecular features of the cytotoxicity of an NHE inhibitor: Evidence of mitochondrial alterations, ROS overproduction and DNA damage
Francesca Aredia
BMC Cancer, 16 (2016)
Isolation and characterization of human breast tumor-derived endothelial cells
Cristina Grange
Oncology Reports, 15(12), 381-386 (2006)
Rashmi Singh et al.
Aquatic toxicology (Amsterdam, Netherlands), 192, 30-39 (2017-09-18)
The current study was aimed to understand the effects of chronic fluoride exposure on fish immune system. African sharp tooth catfish (Clarias gariepinus) were exposed to 73.45mg/L of fluoride corresponding to 1/10 96h LC
Christopher Howell et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 48(9), 1214-1221 (2018-06-21)
Viral infection of the bronchial epithelium disrupts the barrier properties of the epithelium in healthy individuals and those with lung disease. Repair of the bronchial epithelium is dependent of the formation of a provisional fibrin matrix and migration of epithelial
Erik Brauer et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(9), 1801780-1801780 (2019-05-09)
Wound contraction is an ancient survival mechanism of vertebrates that results from tensile forces supporting wound closure. So far, tissue tension was attributed to cellular forces produced by tissue-resident (myo-)fibroblasts alone. However, difficulties in explaining pathological deviations from a successful

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service