Merck
All Photos(1)

D9194

Sigma-Aldrich

Diaminorhodamine-4M AM solution

5 mM in DMSO

Synonym(s):
3′,6′-Bis(dimethylamino)-9-[2-acetomethoxycarbonyl-3-amino-4-(N-methylamino)phenyl]xanthylium iodide, DAR-4M AM solution
EC Number:
NACRES:
NA.32

form

liquid

Quality Level

storage condition

protect from light

concentration

5 mM in DMSO

shipped in

wet ice

storage temp.

−20°C

Application

DAR-4M AM is a cell-permeable dye used for the fluorescent detection of nitric oxide (NO). It can be used to detect NO directly in cell specimens prepared from various sources, including the brain.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service