Merck
All Photos(1)

Documents

EP022639909

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

new
Sign Into View Organizational & Contract Pricing

material

aluminum

feature

RCF 5,005 × g

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 022639909

compatibility

5804/5804R & 5810/5810R

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Rotor F-35-48-17, including 48 steel sleeves and adapters and software upgrade (required for some older models)
  • High-capacity fixed-angle rotor for 15 mL conical and glass tubes
  • Maximum speed: 5,005 × g (5,500 rpm)1)
  • Form inserts for 15 mL conical tubes

1) Please note that a software upgrade will be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information.

Features and Benefits

High-capacity fixed-angle rotor for 15 mL conical and glass tubes
Maximum speed: 5,005 × g (5,500 rpm)1)
Form inserts for 15 mL conical tubes
1) Please note that a software upgrade will be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service