Merck
All Photos(1)

L011800

Sigma-Aldrich

Acetonitrile

Synonym(s):
ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide
Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
PubChem Substance ID:

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

type

for DNA synthesis

product line

Proligo Reagents

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

packaging

pkg of 1 × 18 L L011800-C01-01

impurities

≤10 ppm water content (Karl Fischer)

color

clear Appearance of Solution

refractive index

n/D 1.342-1.346
n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

solubility

water: soluble (completely)

density

0.786 g/mL at 25 °C (lit.)

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service