Merck
All Photos(2)

MAK209

Sigma-Aldrich

Granzyme B Inhibitor Screening Kit

Sufficient for 100 Fluorometric tests

Synonym(s):
GZMB Inhibitor Screening Kit
EC Number:
NACRES:
NA.25

detection method

fluorometric

storage temp.

−20°C

Gene Information

human ... GZMB(3002)
mouse ... GZMB(14939)
rat ... GZMB(171528)

Related Categories

General description

Granzyme B (GZMB; EC 3.4.21.79) is a serine protease stored in the secretory granules of leukocytes. It induces cytotoxic T lymphocyte (CTL)-mediated apoptosis by the activation of caspases and the degradation of structural proteins of the target cells. Granzyme B has an important role in CTL-mediated immune responses and has been implicated in chronic inflammatory diseases such as rheumatoid arthritis, coronary syndrome, and chronic angina pectoris.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the screening of inhibitors of Granzyme B.

Principle

The Granzyme B Inhibitor Screening Kit is a simple and high-throughput assay for screening potential inhibitors of granzyme B. Granzyme B activity is measured by cleaving a synthetic 7-amino-4-trifluoromethylcoumarin (AFC)-based substrate to yield AFC, a fluorescent product (λex = 380/ λem = 500 nm), proportional to the enzymatic activity present. The substrate includes the recognition sequence for granzyme B.

Kit Components Only

Product No.
Description

  • Granzyme B Assay Buffer

  • Granzyme B Substrate

  • Granzyme B Enzyme

  • Inhibitor Control, Ac-IEPD-CHO

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service