20566

Supelco

Leak-Tec® Leak Detector

EC Number:
PubChem Substance ID:

packaging

pkg of 283 g (pressurized can)

application(s)

gas chromatography (GC): suitable

General description

Flows in the air leakage rate can overload the vacuum pump and trigger a process to shut down. Any leakage into the vacuum system can be isolated and eliminated by engaging sensitive and proper leak-detection methods.

Application

Leak-Tec leak detector spray may be used to check for leaks in the optical fiber sensor system prior to the analysis period to determine exhaled organic compounds.

Legal Information

Leak-Tec is a registered trademark of American Gas and Chemical Co., Ltd.

Certificate of Analysis

Certificate of Origin

John J. McKetta Jr.
Encyclopedia of Chemical Processing and Design (1997)
Lurdes I B Silva et al.
Talanta, 83(5), 1586-1594 (2011-01-18)
Breath analysis constitutes a promising tool in clinical and analytical fields due to its high potential for non-invasive diagnostics of metabolic disorders and monitoring of disease status. An optical fiber (OF) sensor has been developed for determination of volatile organic...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service