Product Name
Biliverdin hydrochloride ≥97.0% (TLC)
Product Number
30891
Product Brand
SIGMA
CAS Number
Molecular Weight
619.11
Test
Specification
APPEARANCE (COLOR)Light Green to Very Dark Green
APPEARANCE (FORM)Powder or Crystals
PURITY (TLC AREA %)≥ 97.0 %
CARBON CONTENT64.02 % (THEORY)
HYDROGEN CONTENT5.70 % (THEORY)
NITROGEN CONTENT9.05 % (THEORY)
INFRARED SPECTRUMCONFORMS TO STRUCTURE