Product Name
Dispase® II protease
Product Number
d4693
Product Brand
sigma
CAS Number
42613-33-2
Molecular Weight
Test
Specification
APPEARANCE (COLOR)White to Beige and Faint Brown to Brown
APPEARANCE (FORM)Powder
ENZYMATIC ACTIVITY≥ 0.50 U/mg
UNIT DEFINITIONONE UNIT WILL HYDROLYZE CASEIN TO PRODUCE COLOR EQUIVALENT TO 1.0 UMOLE OF TYROSINE PER MINUTE AT PH 7.5 AT 37 DEG C (COLOR PER FOLIN CIOCALTEU REAGENT)