Merck
1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt solution, 10 mg/mL in chloroform, ≥99%

1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt solution, 10 mg/mL in chloroform, ≥99%
Synonyms:
(9Z)-9-Octadecenoic acid 1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester monosodium salt, 1,2-Di(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, L-α-Phosphatidyl-DL-glycerol, dioleoyl, L-α-Phosphatidyl-DL-glycerol, dioleoyl sodium salt, DOPG-Na, PG(18:1(9Z)/18:1(9Z))
Linear Formula:
C42H78O10PNa
CAS Number:
Molecular Weight:
797.03
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt, ≥98.0% (TLC)
1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt, solution, 10 mg/mL in chloroform, ≥99%