Merck
(S)-(+)-Glutamine for synthesis

L-Glutamine

(S)-(+)-Glutamine for synthesis
Synonyms:
(S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide, L-Glutamine
Linear Formula:
H2NCOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.14

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Analyte Suitability
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Special Grade
Technique