Merck
D-Mannitol NIST® SRM® 920

D-Mannitol

D-Mannitol NIST® SRM® 920
Synonyms:
Mannite, D-Mannitol, Parteck Mannitol
Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
D-Mannitol, ≥98%
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
D-Mannitol, ACS reagent
D-Mannitol, meets EP, FCC, USP testing specifications
D-Mannitol, BioXtra, ≥98% (HPLC)
D-Mannitol, suitable for plant cell culture
D-Mannitol, tested according to Ph. Eur.
D-Mannitol, European Pharmacopoeia (EP) Reference Standard
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
D-Mannitol, United States Pharmacopeia (USP) Reference Standard
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
D-Mannitol, EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP
D-Mannitol, EMPROVE® ESSENTIAL, Ph. Eur., ChP, JP, USP, E 421
D-Mannitol, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP, FCC, E 421
D-Mannitol, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP, E 421
D-Mannitol, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP, E 421
D-Mannitol
D-Mannitol, fine powder, EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, USP, FCC, E 421
D-Mannitol, for the determination of boric acid
D-Mannitol, ≥99.9999% (metals basis), for boron determination
D-Mannitol, suitable for microbiology
D-Mannitol, EMPROVE® EXPERT, Ph. Eur., BP, JP, USP, E 421
D-Mannitol, British Pharmacopoeia (BP) Reference Standard
D-Mannitol, NIST® SRM® 920