Merck
Sodium dodecyl sulfate for ion pair chromatography LiChropur™

Sodium dodecyl sulfate

Sodium dodecyl sulfate for ion pair chromatography LiChropur™
Synonyms:
Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate, Sodium dodecyl sulfate, OmniPur® Sodium Dodecyl Sulfate, Sodium n-Dodecyl Sulfate, Sodium n-Dodecyl Sulfate, Molecular Biology Grade, Sodium n-Dodecyl Sulfate, High Purity, Dodecyl hydrogen sulfate sodium salt
Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-20 of 27
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2
Page 1 of 2