Merck
Phenol, Molecular Biology Grade

Phenol

Phenol, Molecular Biology Grade
Synonyms:
Hydroxybenzene, Phenol solution, Phenol
Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11

Products

Product Category
Brand
Boiling Point (°C)
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Microorganism Suitability
Organoleptic Class
Special Grade
Technique