CH
EN
Applications
Products
Services
Support
Home
Search Results
ABTS
™
Chromophore, Diammonium Salt
(2)
ABTS
™
Chromophore, Diammonium Salt
Synonyms:
ABTS
™
Chromophore, Diammonium Salt
Empirical Formula (Hill Notation):
C
18
H
18
N
4
O
6
S
4
· (NH
3
)
2
CAS Number:
30931-67-0
Molecular Weight:
548.68
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
194437
ABTS
™
Chromophore, Diammonium Salt
Expand
View Pricing
194430
ABTS
™
Chromophore, Diammonium Salt
Expand
View Pricing