Merck
Acetonitrile with 0.1% (v/v) Formic acid hypergrade for LC-MS LiChrosolv®

Acetonitrile with 0.1% (v/v) Formic acid

Acetonitrile with 0.1% (v/v) Formic acid hypergrade for LC-MS LiChrosolv®
Synonyms:
Acetonitrile solution
Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Acetonitrile with 0.1% (v/v) Formic acid, hypergrade for LC-MS LiChrosolv®