CH
EN
Applications
Products
Services
Support
Home
Search Results
Acetonitrile with 0.1% (v:v) trifluoroacetic acid
(2)
Acetonitrile with 0.1% (v:v) trifluoroacetic acid
Synonyms:
Acetonitrile solution
Linear Formula:
CH
3
CN
CAS Number:
75-05-8
Molecular Weight:
41.05
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
4.80448
Acetonitrile with 0.1% (v:v) trifluoroacetic acid, for liquid chromatography LiChrosolv
®
Expand
View Pricing
1.59014
Acetonitrile with 0.1% (v:v) trifluoroacetic acid, hypergrade for LC-MS LiChrosolv
®
Expand
View Pricing