Skip to Content
Products
Cart
0
CH
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Octylbenzene
Synonym(s):
octylbenzene
Linear Formula:
C14H22
CAS Number:
2189-60-8
Molecular Weight:
190.33
Products
Brand
AiFChem (an XtalPi company)
BLD Pharmatech
Fluorochem (Preferred Partner)
Physical Form
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
BL3H160B966E
Octylbenzene,
Expand
View Pricing
XPIH9BCFFEDA
Octylbenzene,
Expand
View Pricing
FLUH99C85F2E
Octylbenzene,
Expand
View Pricing
An unknown error has occured.