Merck
E-Toxate™ Water endotoxin, free

E-Toxate Water

E-Toxate™ Water endotoxin, free
Synonyms:
Water
Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
E-Toxate Water, endotoxin, free