Products
CH
EN
Products
Applications
Products
Services
Support
Home
Search Results
Mowiol
®
56-98
(1)
Mowiol
®
56-98
Synonyms:
Poly(vinyl alcohol)
Linear Formula:
[-CH
2
CHOH-]
n
CAS Number:
9002-89-5
EC Number:
209-183-3
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
10851
Mowiol
®
56-98, M
w
~195,000
Expand
View Pricing