CH
EN
Applications
Products
Services
Support
Home
Search Results
Phenol, Molecular Biology Grade - CAS 108-95-2 - Calbiochem
(1)
Phenol, Molecular Biology Grade - CAS 108-95-2 - Calbiochem
Synonyms:
Phenol, Hydroxybenzene
Linear Formula:
C
6
H
5
OH
CAS Number:
108-95-2
Molecular Weight:
94.11
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
516724-M
Phenol, Molecular Biology Grade - CAS 108-95-2 - Calbiochem
Expand
View Pricing