Merck
  • Conformations of a metastable SH3 domain characterized by smFRET and an excluded-volume polymer model.
Conformations of a metastable SH3 domain characterized by smFRET and an excluded-volume polymer model.
Biophysical Journal (2016)
Mazouchi A, et al.