Merck
  • [Interleukin 7 and its receptor promote cell proliferation and induce lymphangiogenesis in non-small cell lung cancer].
[Interleukin 7 and its receptor promote cell proliferation and induce lymphangiogenesis in non-small cell lung cancer].
Zhonghua bing li xue za zhi = Chinese journal of pathology (2012-11-20)
Jian Ming, Qing-fu Zhang, Yan-duo Jiang, Guo-cheng Jiang, Xue-shan Qiu
ABSTRACT
To study the mechanism of interleukin 7/interleukin 7 receptor (IL-7/IL-7R) in promoting cell proliferation and inducing lymphangiogenesis of non-small cell lung cancer (NSCLC) in vivo and in vitro. Immunohistochemical study for IL-7, IL-7R, cyclin D1 and vascular endothelial growth factor-D (VEGF-D) was carried out in NSCLC tissues from 95 patients. The relationship between IL-7/IL-7R expression and various parameters was analyzed. The mechanism of IL-7/IL-7R in promoting cell proliferation and inducing lymphangiogenesis was studied by methylthiazolyldiphenyl-tetrazolium bromide, fluorescence-activated cell sorting, reverse transcriptase-PCR, Western blot, co-immunoprecipitation, chromatin immunoprecipitation and nude mice experiments with xenograft tumors. IL-7 (63.2%, 60/95), IL-7R (61.1%, 58/95), cyclin D1 (52.6%, 50/95) and VEGF-D (58.9%, 56/95) showed that high level of expression in NSCLC. IL-7/IL-7R over-expression correlated with cyclin D1 expression (P < 0.01, P < 0.01), VEGF-D expression (P < 0.01, P < 0.01), increased lymphovascular density (P = 0.005, P = 0.013), advanced clinical stage (P = 0.008, P = 0.005) and presence of lymph node metastasis (P < 0.01, P < 0.01). IL-7/IL-7R could promote proliferation of A549 cell, increase cyclin D1 and VEGF-D expression, and enhance c-Fos/c-Jun expression and phosphorylation, resulting in formation of heterodimer. Furthermore, IL-7/IL-7R could induce binding of c-Fos/c-Jun to cyclin D1/VEGF-D promoters and regulate their transcription. IL-7/IL-7R could also promote proliferation and lymphangiogenesis of lung cancer xenograft tumors. IL-7/IL-7R promotes c-Fos/c-Jun expression and activity in NSCLC. This further facilitates cyclin D1 expression and accelerates proliferation of cells and VEGF-D-induced lymphovascular formation.