Merck
  • Epithelial-mesenchymal transition (EMT) beyond EGFR mutations per se is a common mechanism for acquired resistance to EGFR TKI.
Epithelial-mesenchymal transition (EMT) beyond EGFR mutations per se is a common mechanism for acquired resistance to EGFR TKI.
Oncogene (2018-08-17)
Chien-Hui Weng, Li-Yu Chen, Yu-Chin Lin, Jin-Yuan Shih, Yun-Chieh Lin, Ruo-Yu Tseng, An-Chieh Chiu, Yu-Hsuan Yeh, Chi Liu, Yi-Ting Lin, Jim-Min Fang, Ching-Chow Chen
ABSTRACT
Epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKI) is a major advance in treating NSCLC with EGFR-activating mutations. However, acquired resistance, due partially to secondary mutations limits their use. Here we report that NSCLC cells with acquired resistance to gefitinib or osimertinib (AZD9291) exhibit EMT features, with a decrease in E-cadherin, and increases in vimentin and stemness, without possessing any EGFR secondary mutations. Knockdown of E-cadherin in parental cells increased gefitinib resistance and stemness, while knockdown of vimentin in resistant cells resulted in opposite effects. Src activation and Hakai upregulation were found in gefitinib-resistant cells. Knockdown of Hakai elevated E-cadherin expression, attenuated stemness, and resensitized the cells to gefitinib. Clinical cancer specimens with acquired gefitinib resistance also showed a decrease in E-cadherin and an increase in Hakai expression. The dual HDAC and HMGR inhibitor JMF3086 inhibited the Src/Hakai and Hakai/E-cadherin interaction to reverse E-cadherin expression, and attenuated vimentin and stemness to restore gefitinib sensitivity. The EMT features of AZD9291-resistant H1975 cells were related to the upregulation of Zeb1. Both gefitinib and AZD9291 sensitivity was restored by JMF3086 through reversing EMT. Our study not only revealed a common mechanism of EMT in both gefitinib and AZD9291 resistance beyond EGFR mutations per se, but also provides a new strategy to overcome it.
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA