  • Search Results
  • Hypermethylation of Secreted Frizzled Related Protein 1 gene promoter in different astrocytoma grades.
Hypermethylation of Secreted Frizzled Related Protein 1 gene promoter in different astrocytoma grades.
Croatian medical journal (2018-11-06)
Anja Kafka, Valentina Karin-Kujundžić, Ljiljana Šerman, Anja Bukovac, Niko Njirić, Antonia Jakovčević, Nives Pećina-Šlaus
ABSTRACT
To identify the involvement of Secreted Frizzled Related Protein 1 (SFRP1) promoter hypermethylation in different malignancy grades of astrocytoma and assess its association with beta-catenin, lymphoid-enhancer factor 1, and T-cell factor 1. Twenty-six astrocytoma samples were collected from 2008-2015. Promoter hypermethylation was evaluated by methylation-specific polymerase-chain-reaction and protein expression by immunohistochemistry and stereological analysis. The staining intensity was scored by comparing immunoreactivity with normal tissue and by using 10% and 50% cut-offs. SFRP1 promoter methylation was found in 32% of astrocytomas. The number of hypermethylated samples increased in higher astrocytoma grades and was the highest in glioblastoma (P=0.042 compared to other astrocytoma grades). There was 45.8% of samples with the lack of or weak expression of SFRP1 protein and 29.2% with strong expression. Samples with methylated promoter expressed significantly less SFRP1 than samples with unmethylated promoter (P=0.031). Beta-catenin expression levels were elevated. Yet, glioblastomas with unmethylated SFRP1 promoter had significantly less beta-catenin (P=0.033). Strong expression of lymphoid-enhancer factor 1 was associated to higher astrocytoma grades (P=0.006). SFRP1 gene was epigenetically silenced in glioblastomas when compared to low astrocytoma grades, which may suggest that the lack of its protein is involved in astrocytoma progression.
Sigma-Aldrich
Anti-ATRX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution