Merck
  • Two Faces of CwlM, an Essential PknB Substrate, in Mycobacterium tuberculosis.
Two Faces of CwlM, an Essential PknB Substrate, in Mycobacterium tuberculosis.
Cell reports (2018-10-04)
Obolbek Turapov, Francesca Forti, Baleegh Kadhim, Daniela Ghisotti, Jad Sassine, Anna Straatman-Iwanowska, Andrew R Bottrill, Patrick J Moynihan, Russell Wallis, Philippe Barthe, Martin Cohen-Gonsaud, Paul Ajuh, Waldemar Vollmer, Galina V Mukamolova
ABSTRACT
Tuberculosis claims >1 million lives annually, and its causative agent Mycobacterium tuberculosis is a highly successful pathogen. Protein kinase B (PknB) is reported to be critical for mycobacterial growth. Here, we demonstrate that PknB-depleted M. tuberculosis can replicate normally and can synthesize peptidoglycan in an osmoprotective medium. Comparative phosphoproteomics of PknB-producing and PknB-depleted mycobacteria identify CwlM, an essential regulator of peptidoglycan synthesis, as a major PknB substrate. Our complementation studies of a cwlM mutant of M. tuberculosis support CwlM phosphorylation as a likely molecular basis for PknB being essential for mycobacterial growth. We demonstrate that growing mycobacteria produce two forms of CwlM: a non-phosphorylated membrane-associated form and a PknB-phosphorylated cytoplasmic form. Furthermore, we show that the partner proteins for the phosphorylated and non-phosphorylated forms of CwlM are FhaA, a fork head-associated domain protein, and MurJ, a proposed lipid II flippase, respectively. From our results, we propose a model in which CwlM potentially regulates both the biosynthesis of peptidoglycan precursors and their transport across the cytoplasmic membrane.
Sigma-Aldrich
Cyanogen bromide-activated Agarose, lyophilized powder
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid
Roche
cOmplete ULTRA Tablets, Mini, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Sigma-Aldrich
GenElute Plasmid Miniprep Kit, sufficient for 350 purifications
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution