CH
EN
Applications
Products
Services
Support
Home
Search Results
Vitamin C status and cognitive function: A systematic review
Vitamin C status and cognitive function: A systematic review
Nutrients (2017)
Travica N, et al.