CH
EN
Applications
Products
Services
Support
Home
Search Results
Essential amino acid ratios and mTOR affect lipogenic gene networks and miRNA expression in bovine mammary epithelial cells
Essential amino acid ratios and mTOR affect lipogenic gene networks and miRNA expression in bovine mammary epithelial cells
Journal of Animal Science and Biotechnology (2016)
Li S, et al.