  • Search Results
  • Dietary curcumin supplement influence on growth, immunity, antioxidant status, and resistance to Aeromonas hydrophila in Oreochromis niloticus
Dietary curcumin supplement influence on growth, immunity, antioxidant status, and resistance to Aeromonas hydrophila in Oreochromis niloticus
Aquaculture (Amsterdam, Netherlands) (2017)
Mahmoud HK, et al.