CH
EN
Applications
Products
Services
Support
Home
Search Results
In vivo imaging of early signs of dopaminergic neuronal death in an animal model of Parkinson's disease
In vivo imaging of early signs of dopaminergic neuronal death in an animal model of Parkinson's disease
Neurobiology of Disease (2018)
Rizzi N, et al.